Would you like to log in again?
FALL LIMITER WITH 17D-2 CARABINER 6-FT
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Personal Fall limiter, Turbolite, With 17D-2 Carabinar, Unit Connector: Aluminum Carabiner, Lanyard End Connector: Aluminum Rebar Hook, 3,600 lb, Size: 6ft
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FALL LIMITER WITH 17D-2 CARABINER 6-FT 76452-410 MFL-12-Z7/6FT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty