Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Huma OsteoGelatin Medium Bloom Gelatin, Lyophilized, Humabiologics

Supplier: Humabiologics

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BGML-250MG BGML-1G BGML-500MG BGML-100MG BGML-5G
76452-636EA 1637.19 USD InStock
76452-636 76452-634 76452-632 76452-630 76452-628
Huma OsteoGelatin Medium Bloom Gelatin, Lyophilized, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human bone gelatin for hydrogel and bioink applications.

  • Native human bone gelatin
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Lyophilized for tailored 3D hydrogel or bioink applications
  • Supports various cell attachments
  • Bloom strength is 175 to 225 g

Huma OsteoGelatin is isolated from human bone sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. Huma OsteoGelatin has undergone a proprietary manufacturing method to extract human gelatin from bone. Huma OsteoGelatin bone-derived gelatin is supplied lyophilized for hydrogel, bioink, or cell culture applications.

Huma OsteoGelatin addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of Huma OsteoGelatin has undergone extensive testing to meet high safety and quality requirements for translational research needs.

All tissue has been tested for safety and found negative for communicable diseases and infectious agents.

Ordering information: Huma OsteoGelatin Medium Bloom is available in 100, 250, 500 mg and 1 and 5 g (5×1 g).

Caution: Huma OsteoGelatin is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty