Would you like to log in again?
HumaMatrix Coat Native Human-Derived ECM, 1 mg/ml Solution, Humabiologics
Supplier: Humabiologics
HMSS-25ML HMSS-10ML
76452-672EA 286.49 USD InStock
76452-672 76452-670
HumaMatrix Coat Native Human-Derived ECM, 1 mg/ml Solution, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human extracellular matrix (ECM) for cell culture coating.
  • Native human extracellular matrix (ECM) for thin coating
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Ready to use
  • Supports various cell attachments
HumaMatrix Coat is isolated from nutrient-rich human tissue sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaMatrix Coat has undergone a proprietary manufacturing method to preserve native matrix proteins and growth factors. HumaMatrix Coat is supplied at 1 mg/ml for cell culture applications and can be diluted for custom applications.
HumaMatrix Coat addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaMatrix Coat has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
Ordering information: HumaMatrix Coat is available at 1 mg/ml in 10 (2×5 ml) and 25 ml.
Caution: HumaMatrix Coat is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty