Would you like to log in again?
GLOVES LATEX MICRO SZ5.5 CS200 1PAIR
Supplier: Ansell Healthcare
Surgical Gloves, Material: Natural Rubber Latex, Powder-Free, Color Brown, Thickness: Micro, Cuff Style Beaded with Surefit Technology, External Surface: Smooth with Micro-Textured Finish, Internal: Polymer Coated with Dermashield Technology, exceptional dexterity and tactile sensitivity, Size: 5.5
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES LATEX MICRO SZ5.5 CS200 1PAIR 76452-794 5787000 Case of 200 (1/Itm/Bx) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty