Would you like to log in again?
Encore 85 Latex Surgical Gloves, Sterile, Powder-Free, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Encore Latex
5785005 5785001 5785001 5785003 5785006 5785006 5785000 5785000 5785005 5785007 5785003 5785004 5785002 5785004 5785001 5785007
76452-812CS 803.26 USD InStock
76452-812 76452-824 76452-810 76452-822 76452-820 CA32918-093 CA32918-094 CA32918-095 CA32918-096 CA32918-097 CA32918-090 CA32918-091 76452-818 32918-091 76452-816 76452-814
Encore 85 Latex Surgical Gloves, Sterile, Powder-Free, Ansell
Gloves
This natural rubber latex, powder-free surgical glove has a textured finish for superior grip and handling, and its low allergen levels help reduce the risk of latex sensitization in healthcare workers and patients.
  • Low allergen levels
  • Powder-free
  • Textured finish for superior grip
  • Great tactile sensitivity
A textured finish means unequaled reliability when using instruments that require greater gripping capability. The thickness of these gloves is optimized for the best balance of protection and tactile sensitivity, all while allowing for a comfortable fit during the majority of surgical procedures.
Certifications: Manufacturing Facility Registered to ISO 9001 and 13485 (The latter supersedes EN 46001). Contains 50 micrograms or less of total water-extractable protein per gram.
Packaging: Packaged 200 pairs; 50 pairs/dispenser; 4 boxes/case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty