Would you like to log in again?
VWR® SMS Coveralls, Made in USA
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
611-7XL 616-6XL 611-M 604-XL 601-3XL 603-2XL 613-4XL 616-XL 614-3XL 613-M 602-XL 611-4XL 616-M 616-4XL 616-5XL 603-3XL 603-5XL 602-2XL 614-6XL 614-L 611-6XL 603-XL 601-XL 601-6XL 603-S 617-6XL 611-L 604-3XL 617-2XL 612-M 604-S 614-2XL 601-2XL 616-3XL 614-7XL 603-4XL 602-7XL 601-4XL 604-4XL 613-7XL 614-M 601-7XL 611-S 613-2XL 603-L 616-2XL 604-L 611-XL 613-S 613-6XL 612-4XL 617-M 616-7XL 602-6XL 612-2XL 604-6XL 612-6XL 601-S 603-7XL 617-3XL 602-3XL 612-S 603-6XL 617-S 602-5XL 614-5XL 616-S 617-L 614-4XL 617-5XL 612-3XL 602-L 604-5XL 617-XL 602-M 617-7XL 613-XL 617-4XL 616-L 614-XL 604-7XL 614-S 604-2XL 601-M 613-3XL 604-M 613-5XL 612-L 612-7XL 602-4XL 612-5XL 611-5XL 602-S 611-2XL 613-L 601-5XL 601-L 603-M 612-XL 611-3XL
76470-516CS 725.02 USD InStock
76470-516 76456-916 76470-514 76456-914 76456-912 76470-518 76456-910 76456-998 76470-512 76470-510 76456-918 76456-996 76456-994 76456-992 76456-990 76456-904 76470-526 76456-902 76470-524 76456-988 76456-900 76456-986 76470-528 76456-908 76470-522 76456-906 76470-520 76456-984 76456-982 76456-980 76470-538 76470-536 76456-978 76456-976 76470-530 76470-496 76470-494 76470-534 76470-532 76470-498 76456-974 76456-972 76456-970 76470-548 76456-968 76470-546 76456-966 76456-964 76457-002 76470-540 76470-544 76457-000 76470-542 76456-962 76456-960 76470-558 76456-958 76456-956 76456-954 76470-552 76470-550 76470-556 76470-554 76457-138 76456-952 76456-950 76456-948 76456-946 76470-568 76457-140 76456-944 76456-942 76470-562 76457-146 76470-560 76457-148 76457-142 76470-566 76457-144 76470-564 76470-570 76456-940 76456-938 76457-150 76456-936 76456-934 76456-932 76470-572 76457-152 76456-930 76456-926 76470-504 76456-924 76470-502 76456-922 76470-508 76456-920 76470-506 76470-500 76456-928
VWR® SMS Coveralls, Made in USA
Disposable Garments
VWR® Made in USA SMS Coveralls are an ideal option for single-use needs.
  • 3-layer SMS construction
  • Serged seams
  • Produced with latex-free elastic
  • Made in USA
Coveralls are manufactured from a unique multi-layered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed 25 coveralls per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty