Would you like to log in again?
Whatman™ Nuclepore Hydrophilic Membrane, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Whatman™ Nuclepore™ Nuclepore™
110405 10417018 800309 112104 10418312 10419331 10417301 110602 10418301 10419506 10418812 10419512 10417412 10419539 10417050 10417112 10419501 10417014 10418818 10419318 10417306 10417012 110601 10418739 10417006 111114 10417018 112803 10417106 10417104 110413 10419531 10417012 10417051 10418701 10417004 10417001 10419312 10418301 10417031 110409 10417512 10418401 110605 111705 10419331 10419518 110610 110401 112105 110605 10418506 800308 10417406 800319 110601 112805 10417206 10418306 10418514 10419318 111503 10417401 110614 10417501 10418704 10417001 110603 112805 10418806 10417004 10419518 111103 10419512 110604 112104 112105 112810 10417031 111112 10418718 110602 110604 111703 113502 111101 10417312 10419312 10418312 113313 10417304 10419531 111105 110409 10417051 10417551 800308 10417101 10417506 10417006 10417014 800309 110611 10419339 111101 10419501 800307 800307 10419306 10417450 110608 10418512 113502 112803 10418501 10419539 111103 110613 111705 10417414 111114 10417114 10418731 10419504 111703 10417212 110405 113506 111105 10418706 10419506 10417139 10417309 10418406 10419339 10419306 110603 111503 10418412 10419504 10418712 110410 110401 10417118 111104 10418318 10418306
76457-208PK 363.85 USD InStock
76457-208 76881-928 CA11013-632 CA28157-836 76457-204 76881-926 76457-206 76881-924 28157-666 10034-600 CA28157-790 10034-566 76457-170 76457-172 76332-644 76332-642 CA28158-522 76332-640 76457-178 CA76457-220 CA28158-125 76457-174 76457-176 76457-210 CA76881-928 76881-918 CA76881-926 76881-916 CA76881-924 28157-814 CA28157-666 76457-218 CA76881-922 76332-628 CA28158-806 76332-626 CA76332-636 76332-636 76457-182 76457-184 76332-634 CA76332-640 76332-632 97058-240 76457-180 76332-630 CA76457-198 76457-222 76881-922 28158-125 76881-920 CA28158-136 76457-186 76457-220 CA28158-373 76457-188 CA76881-918 76881-908 CA76881-916 76881-906 10128-880 76457-226 CA97058-238 76881-904 76457-228 76881-902 CA11013-330 CA76332-626 76332-618 76332-616 76332-624 76457-192 76332-622 76457-194 CA76332-632 76332-620 76457-190 CA28158-624 97058-234 76332-584 CA76881-920 76881-910 76332-582 10128-878 CA97058-240 97058-238 76457-196 76332-580 76457-198 CA76881-906 CA76881-904 28157-836 CA76881-902 CA11013-342 76332-608 76332-606 76332-604 CA76332-614 CA76881-908 28157-790 76332-614 76332-578 CA15000-610 76332-612 76332-576 76332-610 76332-574 76332-572 CA76881-910 CA15000-616 CA76332-582 CA97058-234 CA15000-612 28158-067 CA15000-614 CA10128-878 15000-612 CA76332-572 CA28158-840 15000-614 CA10128-880 15000-616 CA11013-324 CA11013-684 CA11013-320 10034-598 CA28158-737 CA28158-453 28158-840 CA28157-814 10034-584 10034-586 11008-586 10034-588 CA11008-586 CA76457-200 10034-572 10034-574 CA11013-302 CA11013-300 10034-570 CA10033-666 28158-668 CA76457-176 76457-200 76457-202 CA28158-158
Whatman™ Nuclepore Hydrophilic Membrane, Whatman products (Cytiva)
Filters Membrane Filters
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
  • Track-etched for highly defined pore size and high pore density
  • Treated with polyvinylpyrrolidone (PVP) to render it hydrophilic
  • Low extractables, minimizing filtration sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat membrane surface for high particle visibility
Cytiva's Whatman Nuclepore hydrophilic membrane filter range is manufactured from high-quality polycarbonate film track-etched (PFTE) membrane for high nominal pore size precision.
These sharply defined cylindrical micropores, combined with high flow rates and chemical and thermal resistance, make Nuclepore filtration membranes suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty