Would you like to log in again?
VWR® Premium Mayo Dissecting Scissors, German Steel
Supplier: Avantor
VWR®
11647-263
76457-344EA 55.76 USD InStock
76457-344
VWR® Premium Mayo Dissecting Scissors, German Steel
Scissors
These high-quality German steel dissecting scissors are versatile and can be used in multiple procedures and settings
  • Large rings for comfort, user control and stability
  • Manufactured from ASTM-standard German stainless steel
  • Anti-glare coating
  • Autoclavable and corrosion-resistant
Top-tier instrument with superior craftsmanship. Product conforms to international quality standards.
Certifications: ISO, GMP
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty