Would you like to log in again?
VWR® Dissecting Scissors with Open Shanks
Supplier: Avantor
VWR®
11647-277 11647-276
76457-370EA 10.08 USD InStock
76457-370 76457-372
VWR® Dissecting Scissors with Open Shanks
Scissors
These high-quality dissecting scissors are primarily used for general, multipurpose cutting and dissection applications.
  • Large rings for comfort, user control and stability
  • Manufactured from AISI 420 stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
Product conforms to international quality standards.
Certifications: ISO, GMP
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty