Would you like to log in again?
VWR® General Purpose Dissecting Scissors
Supplier: VWR
VWR®
11647-278 11647-290 11647-285 11647-281 11647-295 11647-283 11647-287 11647-282 11647-294 11647-286 11647-291 11647-288 11647-292 11647-279 11647-289 11647-293 11647-280 11647-284
76457-406EA 18.84 USD InStock
76457-406 76457-408 76457-402 76457-404 76457-390 76457-380 76457-392 76457-382 76457-398 76457-376 76457-388 76457-400 76457-378 76457-394 76457-384 76457-374 76457-396 76457-386
VWR® General Purpose Dissecting Scissors
Scissors
These high-quality dissecting scissors are primarily used for general and multipurpose cutting, suture removal and dissection applications.
  • Large rings for comfort, user control and stability
  • Manufactured from AISI 420 stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
Product conforms to international quality standards.
Certifications: ISO, GMP
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty