Would you like to log in again?
VWR® Jar Forceps with Serrated Tips
Supplier: VWR
VWR®
11647-298 11647-297 11647-296
76457-414EA 25.04 USD InStock
76457-414 76457-410 76457-412
VWR® Jar Forceps with Serrated Tips
Forceps
These high-quality thumb forceps remove specimens from deep bottles or jars.
  • Manufactured from AISI 400 series stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
For classroom dissecting, field-study, removal of small embedded objects and crafts.
Certifications: ISO, GMP
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty