Would you like to log in again?
VWR® Post-Mortem Autopsy Scalpel Blades
Supplier: Avantor
VWR®
11647-311 11647-304
76457-426PK 111.91 USD InStock
76457-426 76457-440
VWR® Post-Mortem Autopsy Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
These high-quality Post Mortem Autopsy Scalpel Blades are made from surgical carbon steel blades and sharpened to perfection for a super clean cut.
  • Thicker blades reduce breakage
  • Surgical-quality blades
  • Individually sealed and peelable packaging
  • Sterilized with gamma radiation
For post-mortem, autopsy and research use. Fits VWR Post Mortem Scalpel Knives.
Certifications: ISO, GMP
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty