Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Stainless Steel Scalpel Blades
Supplier: VWR
VWR®
11647-332 11647-327 11647-314 11647-330 11647-313 11647-326 11647-312 11647-329 11647-328 11647-331
76457-480PK 88.07 USD InStock
76457-480 76457-470 76457-446 76457-442 76457-476 76457-444 76457-478 76457-482 76457-472 76457-474
VWR® Stainless Steel Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
VWR® Stainless Steel Surgical Scalpel Blades are made from sharp, long lasting surgical stainless steel.
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade, stainless-steel blades
  • Individually sealed with peelable packaging
  • Sterilized with gamma radiation
Sterile. 100 per pack.
Certifications: ISO, GMP
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty