Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Carbon Steel Scalpel Blades
Supplier: VWR
VWR®
11647-319 11647-316 11647-322 11647-324 11647-320 11647-318 11647-315 11647-323 11647-321 11647-317 11647-325
76457-468PK 88.07 USD InStock
76457-468 76457-458 76457-448 76457-464 76457-454 76457-466 76457-456 76457-460 76457-450 76457-462 76457-452
VWR® Carbon Steel Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
VWR® Carbon Steel Scalpel Blades are made from high quality carbon steel and sharpened to perfection.
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade, carbon-steel blades
  • Individually sealed with peelable packaging
  • Sterilized with gamma radiation
Sterile. 100 per pack.
Certifications: ISO, GMP
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty