Would you like to log in again?
KIT VIAL /CAP AMBER 300UL 100 SETS KT100
Supplier: MicroSolv Technology Corp
Vial & Cap Kit, Includes 300ul, Fused Insert, Screw Top, Amber Autosampler Vials and 100 EA matching Screw Caps with White Silicone Rubber / Red PTFE Soft-Guard Septa, bonded in Blue Screw Caps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT VIAL /CAP AMBER 300UL 100 SETS KT100 76457-546 9532S-E-1AP Kit of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty