Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Platelet Storage, Incubators and Agitators, FS100, Boekel Scientific
Supplier: Boekel Scientific
FS100
76457-550EA 21938.14 USD InStock
76457-550
Platelet Storage, Incubators and Agitators, FS100, Boekel Scientific
Shakers and Mixers Shaking Incubators
This platelet incubation/agitation system features a thermoelectric temperature control method and has an advanced control system with many automated processes including high low temperature alarm testing, unit temperature calibration and the inventory management system. The inventory management allows for platelet expiration tracking using color coded icons. This easy-to-use tool was designed to decrease loss of platelets due to time expiration.
  • Massive 7" industrial touch screen with intuitive icons
  • Corrosion resistant locking casters for easy unit placement
  • Alarm and standard operation indicator light
  • Onboard thermal printer for keeping records of temperature data
  • Battery backup in the event of a power failure
  • Downloadable event log
  • Gas-less/compressor less heating and cooling design
The FS100 has smooth rolling, lockable casters making it easily to install and relocate. If a blood bank uses a secondary temperature monitoring system they can easily attach a probe using the FS100 probe port and mounting hardware.
These systems also have an integrated thermal printer for printing temperature and event reports. This service free device replaces the old-style chart recorders that are difficult to read and require monthly maintenance. The control systems also have a USB port for downloading unit data and an ethernet port for connection the equipment to a network. Also features RS485 interface.
Delivery information: Supplied with two year warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty