Would you like to log in again?
VWR® Microporous Material Coveralls – Made in USA
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
2614-7XL 2602-3XL 2604-6XL 2616-XL 2602-M 2614-M 2612-4XL 2602-2XL 2614-L 2614-4XL 2612-M 2022-L 2604-L 2616-2XL 2616-6XL 2616-S 2612-5XL 2602-7XL 2602-5XL 2614-6XL 2602-XL 2604-XL 2602-6XL 2616-4XL 2614-2XL 2604-2XL 2612-2XL 2604-M 2616-3XL 2604-5XL 2614-S 2604-4XL 2614-3XL 2604-S 2612-S 2616-5XL 2602-4XL 2612-XL 2604-7XL 2612-3XL 2616-7XL 2616-L 2612-L 2614-5XL 2602-S 2612-7XL 2604-3XL 2614-XL 2616-M 2612-6XL
76457-604CS 530.8 USD InStock
76457-604 76457-648 76457-606 76457-600 76457-644 76457-602 76457-646 76457-608 76470-636 76470-634 76470-638 76457-652 76457-610 76457-654 76470-632 76457-650 76470-630 76457-616 76457-618 76457-612 76457-614 76470-648 76470-646 76470-640 76457-620 76470-644 76470-642 76457-626 76457-628 76457-622 76457-624 76470-658 76470-656 76457-596 76457-630 76470-650 76457-598 76457-632 76470-654 76470-652 76457-638 76457-634 76457-636 76470-668 76470-662 76457-640 76470-660 76457-642 76470-666 76470-664
VWR® Microporous Material Coveralls – Made in USA
Disposable Garments
The VWR® Made in USA PPE line has been expanded to include microporous material coveralls.
  • Microporous, 60 gsm, two-layer material
  • Fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • Hood and Boot options
  • Elastic or open cuff and ankle options
Microporous material coveralls provide increased fluid protection with great comfort. Choose from a variety of styles, including different wrist and ankle options, with or without hoods, boots, or both.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed 25 coveralls per case.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty