Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Microporous Material Lab Coats (Made in USA)
Supplier: Avantor
VWR®
24002-5XL 24006-4XL 24006-M 24002-4XL 24010-L 24005-S 240063XL 24006-S 24002-3XL 24006-7XL 24005-M 24002-L 24002-XL 24002-6XL 24002-S 240053XL 24010-M 240103XL 24010-5XL 24005-5XL 24005-4XL 24006-XL 240052XL 24005-L 240062XL 24006-6XL 24005-XL 24002-7XL 24005-6XL 240102XL 24010-7XL 24002-2XL 24006-L 24010-S 24002-M 24010-6XL 24010-4XL 24010-XL 24005-7XL 24006-5XL
76457-666CS 697.31 USD InStock
76457-666 76457-688 76470-690 76457-700 76457-668 76457-702 76457-690 76470-678 76470-672 76457-696 76457-674 76470-694 76470-670 76470-692 76457-676 76457-698 76457-670 76470-676 76457-692 76470-698 76470-674 76457-694 76470-696 76457-672 76470-680 76457-656 76457-678 76457-658 76470-700 76457-662 76457-684 76470-684 76457-686 76470-682 76457-664 76457-680 76470-688 76470-686 76457-682 76457-660
VWR® Microporous Material Lab Coats (Made in USA)
Disposable Garments
The VWR® Made in USA PPE line has been expanded to include microporous material laboratory coats.
  • Microporous two-layer material, 60 gsm
  • Fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • 3 Pocket or no pocket options
  • Knit or standard collar
  • Open, elastic, or knit cuff
Microporous material laboratory coats offer increased fluid protection with great comfort. Available styles include different wrist, collar, and pocket options.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed in cases of 30 coats.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty