Would you like to log in again?
TUBE SET SHIGEMI D2O GAS-TIGHT LPV 5MM
Supplier: SP Industries
Tube set, LPV Shigemi, D2O, 12mm Bottom, Standard full sets include one outer tube and one inner insert. Gas-Tight sets include one outer tube fused with a complete LPV valve, one inner insert with detachable PTFE Holder and one Positioning rod, Dimension: 5MM
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE SET SHIGEMI D2O GAS-TIGHT LPV 5MM 76458-240 BMS-005J-LPV Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty