Would you like to log in again?
WHEATON® Polypropylene Centrifuge Tubes, DWK Life Sciences
Supplier: DWK Life Sciences
380100 380240 380220
76458-264CS 282.79 USD InStock
76458-264 76458-262 76458-266
WHEATON® Polypropylene Centrifuge Tubes, DWK Life Sciences
Tubes Centrifuge Tubes
Ideal for centrifugation and general liquid handling purposes.
  • Plastic graduated centrifuge tubes manufactured from virgin, medical grade polypropylene
  • Fitted with leakproof, HDPE caps
  • Option of 15 or 50 ml capacity
  • Crisp black printed graduations with large white write on area
  • Supplied sterile by ETO
  • Supplied DNase/RNase-free, non-pyrogenic <0.25 EU/ml
  • Packed in bags
Manufactured from ultra-clear virgin polypropylene, the WHEATON® centrifuge tube range meets the needs of most laboratory users for routine centrifugation applications. Ideal for a wide range of sample handling applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty