Would you like to log in again?
GLOVES NTRL WHT N-ST XL F/DONNING PK100
Supplier: KIMBERLY CLARK CORP
Gloves, Non-Sterile, Nitrile, G5 Cleanroom, 6 Mil, Ambidextrous with high tack, Colour: white, offer textured fingertips, Feature beaded cuffs for easy donning, Double bagged with case liner, recommended for ISO Class 5 or higher cleanroom, to help prevent roll-down, measure 10 in l, Size: xl
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NTRL WHT N-ST XL F/DONNING PK100 76458-658 56868 Case of 1000 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty