Would you like to log in again?
LAB COAT BLUE NON ST W/KNIT CUFF L CS25
Supplier: KIMBERLY CLARK CORP
Lab coat, Colour: Blue, Universal Precautions, made with a protective 3layer SMS fabric and are recommended where there is potential low to moderate exposure to fluids, Offers exceptional resistance to blood strike-through at various levels of exposure, Traditional collar and knit cuffs, Size: Large
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAB COAT BLUE NON ST W/KNIT CUFF L CS25 76458-672 10047 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty