Would you like to log in again?
GLOVES VINYL BEADED CUFF 8 MEDIUM CS1000
Supplier: KIMBERLY CLARK CORP
Gloves, G5 Cleanroom, Non-Sterile, Co-Polymer, Material: Vinyl, beaded cuffs for easy donning, Smooth fingertips, 3 Mil, Ambidextrous, have a smooth exterior finish, Double bagged with case liner, recommended for ISO Class 5 or higher cleanrooms, economical, for tactile sensitivity, Size: 8 medium
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES VINYL BEADED CUFF 8 MEDIUM CS1000 76458-674 61002 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty