Would you like to log in again?
Human Recombinant MMP-9 (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB286106-10UG AB286106-50UG
76458-832EA 655.92 USD InStock
76458-832 76458-834
Human Recombinant MMP-9 (from E. coli)
Proteins and Peptides
Matrix metallopeptidase 9 (MMP-9), also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B, is the mostly studied MMP, due to its fundamental role in cancer biology, autoimmune disease, and other conditions. This enzyme degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP-9 is structurally a multi-domain metalloenzyme, composed of a prodomain, a catalytic domain, a gelatin binding domain, a metal-binding domain, and a carboxyl terminal hemopexin like domain. This active human MMP-9 is composed of the catalytic domain, a gelatin binding domain, and a metal-binding domain (AA 107-457). The protein was expressed in E. coli and purified and refolded using proprietary techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty