Would you like to log in again?
GLOVES EXAM NITRILE BLACK 9.6IN XS BX100
Supplier: Ansell Healthcare
Exam Gloves, 93-732, 1.5 AQL, Material: Nitrile, Color: Black, Chlorinated, Silicone Free, Textured Fingers, Beaded Cuff, Enhanced dexterity and tactility in a thin, yet strong design, Silicone free to ensure advanced product protection, Length: 9.6inch, Size: XS(5.5 - 6)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES EXAM NITRILE BLACK 9.6IN XS BX100 76459-130 93732060 Box of 100 Retrieving
Case of 1000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty