Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Vinyl Examination Gloves
Supplier: Avantor
VWR®
VGPF3001 VGPF3004 VGPF3002 VGPF3003
76459-214CS 221.43 USD InStock
76459-214 76459-216 76459-218 76459-220
VWR® Vinyl Examination Gloves
Gloves
This formulated blend of vinyl provides protection in low risk enviornments while maintaining protection, comfort, and dexterity.
  • Soft fit and feel
  • Latex-free and powder-free
  • DOP and DEHP (phthalate) free
  • Smooth inner coating for easy donning, and to help avoid irritation
  • Applicable for many industries
Suitable alternative for individuals with Type I allergies. Great for a wide range of applications including medical, examination, healthcare, nursing, laboratory, IT, general cleaning, and industrial use.
Meets ASTM D5250 standards for poly-vinyl chloride gloves for medical applications. Meets EN 374 and EN 455 standards. FDA 510k certified and meets food handling requirements.
Packaging: Pack of 100 gloves, case of 10 packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty