Would you like to log in again?
BOTTLE GLASS PTFE LINED CAP QAL 2OZ CS1
Supplier: C&G Containers
Bottle, Straight Side CR 53-400, Material: Glass, Color: clear, with a white Polypropylene plastic PTFE-lined cap, certified by cleaning protocol A1, barcoded bottles, sheets of sample labels QAL, Size: 2oz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOTTLE GLASS PTFE LINED CAP QAL 2OZ CS1 76459-232 QLFS15900241 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty