Would you like to log in again?
Sce PUS1
Supplier: New England Biolabs
M0526S
76459-498EA 85 USD InStock
76459-498
Sce PUS1
Enzymes
Sce Pseudouridine Synthase I (Sce PUS1) is expressed as a 6X His tagged recombinant protein and converts Uridine to Pseudouridine in RNA.
  • Sce PUS1 isomerizes uridine to pseudouridine in predominantly single-stranded RNA in vitro
  • Sce PUS1 is a stand-alone enzyme and does not require an RNA guide or co-factor to modify RNA
Sce PUS1 has a preference for Uridines in single stranded RNA regions over Uridines in double-stranded RNA. Optimal substrate is an unstructured RNA that is 15 nt long or longer. Sequence-specific pseudouridine modification by Sce PUS1 is an alternative to randomly incorporated modified nucleosides by RNA polymerases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty