Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Wilmad-LabGlass Thin-Walled Precision NMR Sample Tubes, 3mm, SP Industries

Supplier: SP Industries
328-PP-8-5 341-PP-7-5
76459-612PK 120.62 USD InStock
76459-612 76459-614
SP Wilmad-LabGlass Thin-Walled Precision NMR Sample Tubes, 3mm, SP Industries
Tubes NMR Tubes
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming.

  • Manufactured in a state-of-the-art ISO 9001:2008 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor
  • Inner surface is resistant to strong acid and base at ambient temperature
  • Can be operated safely at temperatures up to 230 °C, and within a temperature step of 120 °C
  • Rated as hydrolytic class 1
  • Ideal for experiments requiring critical shimming quality (high/ultrahigh field, non-spinning, multi-dimensional, multi-nuclei, DNP experiments and studies involving biological samples)
  • Made from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks

Tight ID and OD tolerance as small as 0.0065 mm accommodates Wilmad inserts.

Note: Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.

Ordering information: Include disposable cap.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty