Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Wilmad-LabGlass Precision NMR Suprasil® (Synthetic Quartz) Tubes, SP Industries

Supplier: SP Industries
507-PP-9SUP-5 528-PP-7SUP-5
76459-620PK 584.5 USD InStock
76459-620 76459-858
SP Wilmad-LabGlass Precision NMR Suprasil® (Synthetic Quartz) Tubes, SP Industries
Tubes NMR Tubes
Wilmad precision NMR Suprasil® (Synthetic Quartz) tubes are manufactured from high purity synthetic fused silica materials with <0.005 ppm Fe₂O₃ and outstanding optical characteristics in the deep UV to the near IR.

  • Transmission rate from 190 to 2600 nm is well over 95% (10 mm thickness) excluding reflection
  • Possesses a similar thermal expansion rate and tensile strength as natural quartz which is 14 times more robust during the cooling/heating process than Type 1 Class A glass
  • Tight Int.Ø and Ext.Ø tolerance as small as 0.0065 mm accommodates Wilmad inserts
  • To maximize SNR, Wilmad Precision NMR Suprasil® Tubes have minimal paramagnetic impurities that would impact shimming
  • Inner surface is resistant to strong acid and base at ambient temperature
  • Safe for experiments at temperatures up to 1300 °C
  • Includes disposable cap

Ideal for photolysis experiments that employ 266 nm light from a Q-Switched laser or 254 nm light emitted by a mercury low pressure lamp with a Schott UG 5 filter, as well as variable temperature experiments that have a temperature step over 120 °C.

Caution: Included caps are not recommended for use with samples dissolved in chloroform-d or acetone-d₆.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty