Would you like to log in again?
COAXIAL SYSTEM COMPLETE 600MHZ 5MM
Supplier: SP Industries
Coaxial System Complete, 600MHz, provides extra protection for toxic sample. It can also be used with a reference standard which is insoluable in the sample or may cause a reaction. There is approximately a 50 um gap between the inner and outer tubes, Ideal for variable temperature, Size: 5 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COAXIAL SYSTEM COMPLETE 600MHZ 5MM 76459-636 516-CC-5 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty