Would you like to log in again?
SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
521-C-1000 521-C-GRN-1000 521-BLK-100 521-ORG-100 521-WHT-1000 521-C-BLU-100 521-ASST-1000 521-PUR-1000 521-C-YLW-1000 521-WHT-100 521-PNK-100 521-AQA-1000 521-GRN-1000 521-C-BLU-1000 521-YLW-100 521-C-GRN-100 521-AQA-100 521-U 521-YLW-1000 521-PNK-1000 521-C-YLW-100 521-FUH-100 521-P-100 521-R 521-ORG-1000 521-ASST-100 521-PUR-100 521-SKY-100 521-RED-100 521-FUH-1000 521-BLK-1000 521-BLU-100 521-SKY-1000 521-GRN-100 521-RED-1000 521-T 521-C-100 521-BLU-1000 521-G-100
76459-654PK 66.24 USD InStock
76459-654 76459-676 76459-656 76459-678 80080-508 76459-658 80080-490 80080-496 80080-498 80080-510 80080-492 80080-494 80080-516 76459-680 80080-518 76459-660 76459-682 80080-512 76459-662 76459-684 80080-514 76459-664 76459-666 76459-668 76459-648 80080-520 80080-504 80080-506 76459-670 80087-496 80080-522 80080-500 76459-672 76459-650 80080-524 80080-502 80087-494 76459-652 76459-674
SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
Closures Test/Sample Tube Closures
Manufactured from high quality polyethylene or ethylene vinyl acetate to ensure a tight seal.
  • Different colors help to track samples
  • Available in large quantities for high throughput laboratories
Caution: Please avoid using disposable NMR tube caps when CDCl₃ serves as the reference solution as the material(s) could be dissolved. For CDCl₃, we recommend PTFE tube caps.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty