Would you like to log in again?
SP Wilmad-LabGlass NMR Tube Caps, PTFE, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WG-1264-10 WG-1264-4 WG-1265-100 WG-1264-5 WG-1264-3
76459-686PK 62.89 USD InStock
76459-686 76459-688 76459-690 76459-692 76459-694
SP Wilmad-LabGlass NMR Tube Caps, PTFE, SP Industries
Closures Test/Sample Tube Closures
The PTFE NMR tube caps are machined to exact specifications with a smaller gyroradius and more homogeneous mass distribution than disposable caps for better spinning stability.
These caps are recommended in experiments at high to ultra high field and experiments using chloroform as reference solution.
Caution: Only to be used with Wilmad precision NMR tubes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty