Would you like to log in again?
SP Wilmad-LabGlass High Pressure Vacuum NMR Tubes, SP Industries
Supplier: SP Industries
524-PV-8 522-PV-7 524-PV-7 522-PV-9
76459-698EA 279.79 USD InStock
76459-698 76459-700 76459-702 76459-696
SP Wilmad-LabGlass High Pressure Vacuum NMR Tubes, SP Industries
Tubes NMR Tubes
This pressure/vacuum tube is a highly reliable NMR tube for medium range pressure (<300 psi) experiments.
  • PV-ANV valve is made of PTFE and all other parts are borosilicate glass
  • Valve is opened simply by turning counterclockwise
Designed to connect to a ¹/₈" metal (stainless steel or brass) vacuum line using SwageLok® fittings or a rubber vacuum hose and a glass connector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty