Would you like to log in again?
SP Wilmad-LabGlass Precision L, S, and X Band EPR Tubes, SP Industries
Supplier: SP Industries
735-PQ-250M 722-PQ-7 706-PQ-9.50 705-PQ-6.25 748-PQ-7 721-PQ-7 715-PQ-250M 701-PQ-7 705-PQ-250M 714-PQ-100M 735-PQ-100M 725-PQ-159M 726-PQ-9.50 734-PQ-8 733-3PQ-200M 702-PQ-7 723-PQ-7 714-PQ-8
76459-984EA 186.99 USD InStock
76459-984 76459-940 76459-974 76459-920 76459-942 76459-910 76459-954 76459-944 76459-912 76459-914 76459-958 76459-948 76459-916 76459-938 76459-980 76459-970 76459-982 76459-960
SP Wilmad-LabGlass Precision L, S, and X Band EPR Tubes, SP Industries
Tubes NMR Tubes
Precision EPR tubes have 10 to 15 times tighter tolerances than standard tubes.
  • Available in Quartz and Suprasil
  • Individually inspected for impurities and quality
  • Includes disposable grade cap
Ideal for experiments that require a constant filing factor or reproducibility on sample volume in the cavity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty