Would you like to log in again?
INSERT 2.19X1.66X9.3X18.6MM 3.2MM ROTOR
Supplier: SP Industries
Insert, for 3.2mm Rotor, designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids, Sample can be sealed into the insert tube by heat-sealing with a torch or applying a small drop of epoxy to the constricted part as shown below, dimension: 2.19x1.66x9.3x18.6mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INSERT 2.19X1.66X9.3X18.6MM 3.2MM ROTOR 76460-026 DWGSK3083-B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty