Would you like to log in again?
SP Wilmad-LabGlass X Band Capillary Tubes, SP Industries
Supplier: SP Industries
Q-1.0X1.2 Q-1.5X1.8 Q-.9X1.1
76460-074PK 177.08 USD InStock
76460-074 76460-072 76460-070
SP Wilmad-LabGlass X Band Capillary Tubes, SP Industries
Tubes NMR Tubes
Wilmad's X-band EPR tubes are made from clear fused quartz with the lowest impurities to guarantee success in detecting subtle spectroscopic details.
  • Available with both ends open
  • Ideal for applications with limited sample size
  • Easy to seal with a hot flame or sealant of your choice
Rigorous testing is done on each tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty