Would you like to log in again?
MASS MULTIPLIER RING PEEK F/V-NS SPINNER
Supplier: SP Industries
Mass Multiplier Ring, Material: PEEK, with 1 or 2 O-Rings at the top, Cost effective compared to the full VT version for sample changers, Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom, for V-NS Style Spinner,
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MASS MULTIPLIER RING PEEK F/V-NS SPINNER 76460-120 UNI-MASS-MULTI Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty