Would you like to log in again?
JET WASHER MULTI TUBE ISO 29/32 JT 3-5MM
Supplier: SP Industries
Jet Washer/Dryer, Multi Tube, Complete, Joint Type: ISO 29/32 Taper Joint, Dimension: 3-5mm Tubes, 5 PTFE coated stainless steel solvent transfer tubes fit 3, 4 and 5 mm NMR/EPR tubes, PTFE solvent cup and tubing make this unit fully resistant to common organic solvents
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
JET WASHER MULTI TUBE ISO 29/32 JT 3-5MM 76460-140 WG-1209-J2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty