Would you like to log in again?
SP Wilmad-LabGlass Pyrex® NMR Capillary Tubes, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
WG-1364-2.5A-203M WG-1365-2 WG-1364-2-203M WG-1365-1.7 WG-1364-2.5A WG-1364-1.7-203M WG-1364-2 WG-1365-2.5A WG-1365-1 WG-1364-1.7 WG-1364-1 WG-1364-1-203M WG-1364-1.7-5
76460-158PK 45.32 USD InStock
76460-158 76460-148 76460-156 76460-168 76460-146 76460-154 76460-166 76460-144 76460-152 76460-164 76460-150 76460-162 76460-160
SP Wilmad-LabGlass Pyrex® NMR Capillary Tubes, SP Industries
Tubes NMR Tubes
Wilmad's capillary tubes are a low-cost solution for small volume NMR measurement.
  • Available with both ends open or sealed at one end
  • Tested at 400 MHz field
Made of ASTM Type 1 Class A glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty