Would you like to log in again?
SP Wilmad-LabGlass NMR Tubes with Bruker® SampleJet® Caps, SP Industries
Supplier: SP Industries
WG-3000-4-SJ WG-1000-7-SJ WG-3000-7-SJ WG-1000-4-SJ
76460-126EA 184.4 USD InStock
76460-126 76460-124 76460-186 76460-190
SP Wilmad-LabGlass NMR Tubes with Bruker® SampleJet® Caps, SP Industries
Tubes NMR Tubes
These Wilmad-LabGlass NMR tubes are supplied with authentic closed Bruker® SampleJet® caps without code.
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
Designed for routine use in most low to mid field NMR spectrometers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty