Would you like to log in again?
SP Wilmad-LabGlass Gas-Tight Septum NMR Tube Caps, SP Industries
Supplier: SP Industries
WG-3891-100 WG-3889-100
76460-194PK 122.18 USD InStock
76460-194 76460-192
SP Wilmad-LabGlass Gas-Tight Septum NMR Tube Caps, SP Industries
Closures Test/Sample Tube Closures
Gas-tight NMR tube caps are ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar (14.5 psi).
Use these caps with Wilmad medium walled and heavy walled NMR tubes for better seal and robustness in variable temperature experiments. Septa allow easy access via syringe needle.
Caution: Only to be used with Wilmad precision NMR tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty