Would you like to log in again?
ELECTROLYTIC ASSEMBLY SUPRASIL F/BRUKER
Supplier: SP Industries
Electrolytic Assembly, Material: Suprasil, Varian, Radiopan, Electrochemistry has employed in many EPR kinetic studies to generate free radicals, offers complete Electrolytic Cell Assemblies with a Suprasil flat cell, For use with: Bruker ST cavity, Dimension: 10x60x0.5mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELECTROLYTIC ASSEMBLY SUPRASIL F/BRUKER 76460-230 WG-810-Q Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty