Would you like to log in again?
DEWAR INSERT VT CFQ 425X75X75MM OAL
Supplier: SP Industries
Transfer Dewar, VT, Material: CFQ, transfer lines are designed for Bruker ER4121 VT systems, The longer (WG-842-MX and WG-842-ML) versions are recommended for ER 4121 VT system in an EPR console with magnet >18 cm diameter, Dimension: 425x75x75mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DEWAR INSERT VT CFQ 425X75X75MM OAL 76460-262 WG-842-MX Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty