Would you like to log in again?
ROD MOUNTING SUPRASIL SINGLE CRYSTAL 4MM
Supplier: SP Industries
Mounting Rod, Material: Suprasil, Single Crystal, manufactured with the highest grade of synthetic quartz to eliminate E center defects and paramagnetic material contaminations, 2 sizes to choose from, Hexagonal rods are available upon request, Dimension: 4x5x1.5x2mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROD MOUNTING SUPRASIL SINGLE CRYSTAL 4MM 76460-272 WG-856-Q Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty