Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Wilmad-LabGlass Rotors and Caps for Bruker® MAS Probes, SP Industries

Supplier: SP Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

JK-601-7181-WH WP-501-4180-SET-2 WP-601-7181 WP-501-4180-SET-5 JK-601-7181LWH WP-501-2180 JK-603-7181 WP-501-4181 WP-603-3182 WP-501-2180-SET1 WP-501-7180-SET-1 WP-501-7180-SET-5 JK-602-4181 JK-604-4181 WP-602-2182 WP-501-4180-SET-1 WP-501-3180-SET1 WP-501-7180 JK-601-7181 WP-501-7180-SET-2 WP-602-3181 JK-601-7181-L WP-602-2181 WP-603-3181 JK-603-4181 WP-601-4181 WP-501-4180 WP-602-3182 JK-601-4181 WP-501-3180
76460-048EA 155.17 USD InStock
76460-048 76460-302 76460-288 76460-046 76460-322 76460-300 76460-044 76460-286 76460-320 76460-042 76460-284 76460-308 76460-306 76460-304 76460-040 76460-282 76460-312 76460-038 76460-310 76460-036 76460-298 76460-034 76460-296 76460-318 76460-316 76460-314 76460-294 76460-050 76460-292 76460-290
SP Wilmad-LabGlass Rotors and Caps for Bruker® MAS Probes, SP Industries
Centrifuges Rotors
MAS-NMR rotor bodies are manufactured from the highest quality Zirconia, Kel-F, Torlon® and Vespel® providing the ultimate solution for analysis of solid samples.

  • MAS rotors and caps are 100% compatible with most solid state NMR spectrometers
  • Thoroughly inspected before and after the precision machining process to ensure there are no material irregularities
  • Spin testing is performed to the highest specified spinning speed, assuring performance without overspinning the rotor
  • Spinning speeds of up to 12 kHz for 7 mm O.D. rotors
  • Some caps are fitted with O-rings for improved sealing
  • Zirconia rotor body has a strength of 1000 MPa, greater than Si₃N₄

Note: 'DB' is the abbreviation for Bruker® 'Double Bearing' style rotor. 'BL' is the abbreviation for Bruker® 'Boden Lager' (Bottom Bearing) style rotor.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty