Would you like to log in again?
Modern Protein A Antibody Column Package (1-Step Purification Protocol), Cytiva
Supplier: Cytiva
29497628 29497628
76460-354EA 288.46 USD InStock
CA76460-354 76460-354
Modern Protein A Antibody Column Package (1-Step Purification Protocol), Cytiva
Chromatography Columns
The Antibody column package includes modern Protein A affinity resin and desalting columns for purification of human antibodies.
  • Contains columns for a 1-step protocol using affinity chromatography, HiTrap MabSelect PrismA 1×1 ml and HiTrap Desalting 1×5 ml
  • HiTrap MabSelect PrismA column is prepacked with resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
  • HiTrap Desalting column is packed with Sephadex G-25 to give you quick pH neutralization of the eluted antibodies
  • Includes instruction with guidance and tips to quickly get started
The package includes a HiTrap column prepacked with MabSelect PrismA resin containing a protein A-derived ligand produced in E. coli. Fermentation and subsequent purification are performed in the absence of animal products. The ligand has been specially engineered to create an affinity resin with enhanced alkali and protease stability. The specificity of binding to the Fc region of IgG is similar to that of conventional Protein A and provides excellent purification in one step.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty