Would you like to log in again?
His 2-Step Purification Protocol Tagged Column Package, Cytiva
Supplier: Cytiva
29497630 29497630
76460-358EA 945.98 USD InStock
CA76460-358 76460-358
His 2-Step Purification Protocol Tagged Column Package, Cytiva
Chromatography Columns
Column package includes affinity chromatography and size exclusion chromatography columns to achieve very high purity of his-tagged proteins using a 2-step purification protocol.
  • Contains a HisTrap HP column 1 ml prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification of his-tagged proteins
  • Final purity achieved with HiPrep 16/60 Sephacryl S-200 HR size exclusion chromatography column, removes the trace of impurities or closely related substances and imidazole
  • Includes instructions with guidance and tips showing how to get started
Ni Sepharose HP affinity resin consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with nickel, which selectively retains proteins with exposed histidine groups. Ni Sepharose HP has a binding capacity that exceeds 40 mg/ml, resulting in high protein recovery.
The size exclusion column is prepacked with Sephacryl S-200 HR chromatography resin, which has a broad fractionation range of Mr 5000 to ~ 250000 for globular proteins. The high sample capacity of the HiPrep column.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty