Would you like to log in again?
Untagged Chromatography Column Package, Cytiva
Supplier: Cytiva
29497631 29497631
76460-360EA 1029.58 USD InStock
CA76460-360 76460-360
Untagged Chromatography Column Package, Cytiva
Chromatography Columns
The Untagged column package includes cation/anion exchange and size exclusion chromatography columns for purification of untagged/native proteins. The columns provided in the package are prepacked with Capto Q ImpRes and Capto SP ImpRes high-performance resins and can be used individually or in combination.
  • Resins provide you with excellent pressure/flow properties and small resin bead sizes to maximize resolution
  • The resins give you high loading capacities over a broad pH range
Final high purity is obtained with HiPrep 16/60 Sephacryl S-200 HR, a size exclusion chromatography column with a broad fractionation range of Mr 5000 to ~250000 for globular proteins. The high sample capacity of the HiPrep column lets you load samples of up to 5 ml.
Delivery information: Package contains HiTrap Capto Q ImpRes 1×1 ml and Capto SP ImpRes 1×1 ml ion exchange (IEX) columns for effective first-step purification. Includes instruction with guidance and tips to get started quickly.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty