Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ME-T Balances with Internal Calibration, Mettler-Toledo®

Supplier: Mettler-Toledo

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ME-T
30216541 30243388 30704481 30216564 30216550 30216594 30216603 30216545 30216540 30216550 30216542 30216554 30704498 30216593 30704479 30216541 30243383 30216599 30243383 30243388 30216605 30216562 30216558 30216560 30216594 30216603 30216605 30216609 30216540 30216547 30216547 30704487 30216567 30216564 30216554 30216609 30216599 30216593 30216542 30216567 30216607 30216562 30216545 30216560 30216558
76460-389EA 4610 USD InStock
76460-389 CA76460-410 76460-401 76460-423 CA76460-398 76460-387 CA76460-396 76460-421 CA76460-394 CA76460-392 CA76460-418 76460-409 CA76460-416 76460-407 CA76460-414 76460-427 76460-405 CA76460-412 76460-403 CA76460-408 CA76460-390 CA76460-388 76460-411 CA76460-420 CA76460-386 76460-399 76558-330 76460-397 76558-332 76460-395 76460-419 CA76460-406 76460-417 CA76460-404 CA76460-426 76460-415 CA76460-402 CA76460-424 76460-413 CA76460-422 CA76460-400 76460-393 76558-326 76460-391 76558-328
ME-T Balances with Internal Calibration, Mettler-Toledo®
Balances and Scales
Mettler-Toledo® ME-T balances are reliable balances for every laboratory that combine essential weighing functions with ease of use to deliver accurate results.

  • Intuitive operation: 4.5" color touchscreen is easy-to-use, providing fast access to built-in applications
  • Overload protection: Metal base with chemically resistant housing
  • Robust performance: An Electromagnetic Force Compensation (EMFC) weighing cell delivers consistently reliable results and provides protection against accidental overload
  • Easy data transfer: Eliminate manual data recording and transfer results via USB, RS232 and optional Bluetooth

The ME-T Standard Level balances offer high-quality construction and solid performance to ensure consistent and precise weighing results.

The metal base housing and a high quality finish make the ME-T balances robust enough for tough environments, while the smooth surfaces make them easy to maintain and clean.

Ten embedded applications including checkweighing, totaling and formulation allow operators to start working right away. Sample IDs and passcode protection make sure settings are not changed by unauthorized personnel. Multiple interfaces support barcode reader, printer and PC connections for easy traceability and data handling. USB and RS232 connectivity along with an optional Bluetooth adapter simplify data upload and transfer.

Certifications: UL listed and CSA certified.

Ordering information: The 0.01 g and 0.1 g models do not include draft shields.

Order Now
 

ORDER

SERVICES


Services: Optional Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are the key to ensuring the balance performs properly from day one. A Mettler-Toledo technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty